EMAIL THIS PAGE TO A FRIEND
T4937 Sigma-Aldrich

Thiothixene

meets USP testing specifications

Synonym: cis-Thiothixene, N,N-Dimethyl-9-[3-(4-methyl-1-piperazinyl)propylidene]thioxanthene-2-sulfonamide

  • CAS Number 3313-26-6

  • Empirical Formula (Hill Notation) C23H29N3O2S2

  • Molecular Weight 443.63

  •  MDL number MFCD00079574

  •  PubChem Substance ID 24900237

Purchase

Properties

Related Categories Analytical/Chromatography, Core Bioreagents, Enzyme Inhibitors, Pharmacopoeia, Pharmacopoeia A-Z,
InChI Key   GFBKORZTTCHDGY-UWVJOHFNSA-N
suitability   meets USP testing specifications
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
WGK Germany 
3
Protocols & Articles

Articles

HPLC Analysis of Dopamine Receptor Antagonists on Ascentis® Phenyl

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Dopamine Receptor Antagonists on Ascentis® Phenyl
Keywords: Chromatography, Clinical, High performance liquid chromatography, Hormones, Neurotransmitters

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?