• USA Home
  • T5766 - Turkey red oil sodium salt

EMAIL THIS PAGE TO A FRIEND
T5766 Sigma-Aldrich

Turkey red oil sodium salt

for microscopy

Synonym: Castor-oil sulfated sodium salt, Sulfated castor oil, Sulforicinolate sodium salt

Purchase

Properties

Related Categories Alkyl Sulfates, Anionic Detergents, Biochemicals and Reagents, Cell Biology, Detergents,
grade   for microscopy
description   anionic
storage temp.   2-8°C

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
Flash Point(F) 
228.2 °F
Flash Point(C) 
109 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?