EMAIL THIS PAGE TO A FRIEND
T5875 Sigma-Aldrich

(±)-α-Lipoamide

analytical standard

Synonym: α-Lipoic amide, DL-α-Lipoamide, DL-6,8-Thioctic acid amide

  • CAS Number 940-69-2

  • Empirical Formula (Hill Notation) C8H15NOS2

  • Molecular Weight 205.34

  •  Beilstein Registry Number 122470

  •  EC Number 221-710-9

  •  MDL number MFCD00005475

  •  PubChem Substance ID 24900322

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Antioxidant Standards, Chemical Structure,
grade   analytical standard
InChI Key   FCCDDURTIIUXBY-UHFFFAOYSA-N
assay   ≥99%
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
storage temp.   −20°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

MSMLS Online Plate Map

Metabolite descriptors included with the software download upon purchase of the product include: Name, Parent CID, molecular formula, molecular weight, CAS, ChEBI, HMDB ID, PubChem Compound and Subst...
Keywords: Mass spectrometry, Metabolites

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?