• USA Home
  • T6166 - Testosterone isocaproate

EMAIL THIS PAGE TO A FRIEND
T6166 Sigma-Aldrich

Testosterone isocaproate

analytical standard

Synonym: 4-Androsten-17β-ol-3-one 17-(4-methylpentanoate), Testosterone 4-methylvalerate

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Chemical Structure, Chromatography,
grade   analytical standard
InChI Key   PPYHLSBUTAPNGT-BKWLFHPQSA-N
form   neat
drug control   USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?