• USA Home
  • T6284 - Trimebutine maleate salt

EMAIL THIS PAGE TO A FRIEND
T6284 Sigma-Aldrich

Trimebutine maleate salt

analytical standard

Synonym: 3,4,5-Trimethoxybenzoic acid 2-(dimethylamino)-2-phenylbutyl ester

Purchase

Properties

Related Categories Additional Drugs, Additional Standards, Analytical Standards, Analytical/Chromatography, Chromatography,
grade   analytical standard
InChI Key   FSRLGULMGJGKGI-BTJKTKAUSA-N
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Packaging

5 g in poly bottle

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
DI0360000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?