• USA Home
  • T9000 - Thrombin from bovine plasma

EMAIL THIS PAGE TO A FRIEND
T9000 Sigma-Aldrich

Thrombin from bovine plasma

buffered aqueous solution, ≥125 NIH units/mg protein (biuret)

Synonym: Factor IIa

Purchase

Description

Analysis Note

Activity is expressed in NIH units obtained by direct comparison to a NIH Thrombin Reference Standard, Lot K.

The NIH assay procedure uses 0.2 mL of diluted plasma (1:1 with saline) as a substrate and 0.1 mL of thrombin sample (stabilized in a 1% buffered albumin solution) based on a modification of the method of Biggs. Only clotting times in the range of 15-25 seconds are used for determining thrombin concentrations.

Biochem/physiol Actions

Serine protease that selectively cleaves Arg-Gly bonds in fibrinogen to form fibrin and fibrinopeptides A and B.

Other Notes

View more information on thrombin at www.sigma-aldrich.com/enzymeexplorer.

Physical form

In 0.05 M phosphate buffer, pH 7.0

Price and Availability


Laboratory Gloves

Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
XO8950000
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

inhibitor

Product #

Description

Add to Cart

D7910 3,4-Dichloroisocoumarin, serine protease inhibitor
D0879 Diisopropylfluorophosphate
L2023 Leupeptin trifluoroacetate salt, ≥90% (HPLC), microbial
A6191 Antipain dihydrochloride from microbial source, protease inhibitor
H7380 Hirudin from leeches, lyophilized powder, 300-1,500 ATU/mg protein (ATU = antithrombin units)
H7016 Hirudin from leeches, lyophilized powder, ≥1,500 ATU/mg protein (ATU = antithrombin units)
A9141 Antithrombin III from bovine plasma, lyophilized powder, 200-400 units/mg protein (E1%/280 = 6.5)
P7626 Phenylmethanesulfonyl fluoride, ≥98.5% (GC)

substrate

Product #

Description

Add to Cart

B9385 Boc-Val-Pro-Arg-7-amido-4-methylcoumarin hydrochloride
S9009 Sar-Pro-Arg p-nitroanilide dihydrochloride, thrombin substrate
B7632 N-Benzoyl-Phe-Val-Arg-p-nitroanilide hydrochloride, protease substrate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?