• USA Home
  • T9135 - Thrombin from human plasma

EMAIL THIS PAGE TO A FRIEND
T9135 Sigma-Aldrich

Thrombin from human plasma

lyophilized powder

Synonym: Factor IIa

Purchase

Description

Analysis Note

The NIH assay procedure uses 0.2 ml diluted plasma (1:1 with saline) as a substrate and 0.1ml of thrombin sample (stabilized in a 1% buffered albumin solution) based on a modification of the method of Biggs. Only clotting times in the range of 15-25 seconds are used for determining thrombin concentrations.

Biochem/physiol Actions

Serine protease that selectively cleaves Arg-Gly bonds in fibrinogen to form fibrin and fibrinopeptides A and B.

General description

Thrombin is the final coagulation protease in regard to hemostasis, promoting both procoagulant and anticoagulant effects. Lyophilized powder containing sucrose, sodium chloride and Tris.

Other Notes

View more information on thrombin at www.sigma-aldrich.com/enzymeexplorer.

Preparation Note

Prepared from Product No. T7009.

Unit Definition

Activity is expressed in NIH units obtained by direct comparison to a NIH Thrombin Reference Standard

Application

Thrombin is used for site specific cleavage of recombinant fusion proteins containing an accessible thrombin recognition site for removal of affinity tags. Thrombin has been used in a study to assess bleeding risk factors associated with argatroban therapy.

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

inhibitor

Product #

Description

Add to Cart

D7910 3,4-Dichloroisocoumarin, serine protease inhibitor
D0879 Diisopropylfluorophosphate
L2023 Leupeptin trifluoroacetate salt, ≥90% (HPLC), microbial
A6191 Antipain dihydrochloride from microbial source, protease inhibitor
H7380 Hirudin from leeches, lyophilized powder, 300-1,500 ATU/mg protein (ATU = antithrombin units)
H7016 Hirudin from leeches, lyophilized powder, ≥1,500 ATU/mg protein (ATU = antithrombin units)
A9141 Antithrombin III from bovine plasma, lyophilized powder, 200-400 units/mg protein (E1%/280 = 6.5)
P7626 Phenylmethanesulfonyl fluoride, ≥98.5% (GC)

substrate

Product #

Description

Add to Cart

B9385 Boc-Val-Pro-Arg-7-amido-4-methylcoumarin hydrochloride
S9009 Sar-Pro-Arg p-nitroanilide dihydrochloride, thrombin substrate
B7632 N-Benzoyl-Phe-Val-Arg-p-nitroanilide hydrochloride, protease substrate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?