EMAIL THIS PAGE TO A FRIEND
V7763 Sigma-Aldrich

Vitamin A (acetate)

meets USP testing specifications

Synonym: Retinol acetate, Retinyl acetate, Vitamin A acetate

  • CAS Number 127-47-9

  • Empirical Formula (Hill Notation) C22H32O2

  • Molecular Weight 328.49

  •  Beilstein Registry Number 1915439

  •  EC Number 204-844-2

  •  MDL number MFCD00019413

  •  PubChem Substance ID 329829063

Purchase

Properties

Related Categories Analytical/Chromatography, Biochemicals and Reagents, Pharmacopoeia, Pharmacopoeia A-Z, Pharmacopoeial Vitamins and Supplements,
InChI Key   QGNJRVVDBSJHIZ-QHLGVNSISA-N
mp   57-58 °C
suitability   meets USP testing specifications
storage temp.   −20°C

Description

Physical form

Dispersion in a gelatin matrix with sucrose, food starch, methylparabin, propylparabin, and potassium sorbate preservative.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
VH6825000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

HPLC Analysis of Vitamins, Fat Soluble (A and E), on SUPELCOSIL™ LC-18

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Vitamins, Fat Soluble (A and E), on SUPELCOSIL™ LC-18
Keywords: Clinical, High performance liquid chromatography, Vitamins

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

R4632 Retinyl acetate, synthetic, crystalline solid or supercooled liquid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?