EMAIL THIS PAGE TO A FRIEND
01726 Sigma

Acrylamido buffer

~0.2 M in 1-propanol, solution, pKa 6.85

Purchase

Properties

Related Categories Acrylamido Buffers for IPGs, Isoelectric Focusing and Two Dimensional Electrophoresis, Molecular Biology, Protein Electrophoresis, Proteomics More...
form   solution
shelf life   limited shelf life
concentration   ~0.2 M in 1-propanol
pKa    6.85
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 1274 3/PG 2
WGK Germany 
3
Flash Point(F) 
72 °F
Flash Point(C) 
22 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?