EMAIL THIS PAGE TO A FRIEND
03961 Sigma

Mucicarmine

for microscopy

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, M, Stains & Dyes, A to Z, Stains and Dyes More...
grade   for microscopy

Description

Application

Mucicarmine is a histochemical that stains mucins pink. Originally used for the detection of capsular material in fungi, Cryptococcus neoformans, mucicarmine is now used to detect mucin secretions produced by a variety of epithelial and connective tissue cells, especially during certain inflammations and in certain intestinal carcinomas.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?