• USA Home
  • 04088 - (2S,3S)-trans-3-Phenyl-2-oxiranylmethyl 4-nitrophenyl carbonate

EMAIL THIS PAGE TO A FRIEND
04088 Sigma

(2S,3S)-trans-3-Phenyl-2-oxiranylmethyl 4-nitrophenyl carbonate

≥98.0% (NMR)

Synonym: (2S,3S)-2,3-Epoxy-3-phenylpropyl 4-nitrophenyl carbonate, (S)-NEPC, 4-Nitrophenyl (2S,3S)-2,3-epoxy-3-phenylpropyl carbonate

Purchase

Properties

Related Categories Biochemicals and Reagents, Enzyme Substrates, Enzymes, Inhibitors, and Substrates, Epoxy-Hydrolase, Substrates by Enzyme More...
InChI Key   NIEXHSMFFPOGRX-GJZGRUSLSA-N
assay   ≥98.0% (NMR)
optical activity   [α]20/D −60±3°, c = 1% in methylene chloride
mp   69-72 °C
storage temp.   2-8°C

Description

Application

suitable as substrate for epoxide hydrolase

Other Notes

Substrate for cytosolic epoxide hydrolase

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?