• USA Home
  • 04250 - Ethylmagnesium chloride solution

EMAIL THIS PAGE TO A FRIEND
04250 Sigma

Ethylmagnesium chloride solution

technical, ~2.8 M in THF

  • CAS Number 2386-64-3

  • Linear Formula CH3CH2MgCl

  • Molecular Weight 88.82

  •  Beilstein Registry Number 3647886

  •  MDL number MFCD00000472

  •  PubChem Substance ID 329747898

Purchase

Properties

Related Categories Acids & Bases, Alkyl, Bases, Chemical Synthesis, Grignard Reagents,
InChI Key   ZWNJJACKPNTXHY-UHFFFAOYSA-M
grade   technical
concentration   ~2.8 M in THF
density   1.00 g/mL at 20 °C

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
May form explosive peroxides., Reacts violently with water.
RIDADR 
UN 3399A 4.3(3) / PGI
WGK Germany 
1
Flash Point(F) 
1.4 °F
Flash Point(C) 
-17 °C

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Related Products

replaced by

Product #

Description

Add to Cart

303828 Ethylmagnesium chloride solution, 2.0 M in THF

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?