EMAIL THIS PAGE TO A FRIEND
04691 Sigma

NTA - Tracy 690

suitable for fluorescence, ≥80% (HPCE)

Synonym: Nα, Nα-Bis(carboxymethyl)-L-lysine Nickel(II) complex - Tracy 690 conjugate, Tracy 690 - Nitrilotriacetic acid, complexed to Ni2+ - ion

Purchase

Properties

Related Categories Biochemicals and Reagents, Fluorescent Conjugates, Fluorescent NTA Conjugates, Fluorescent Probes, Labels, Particles and Stains
assay   ≥80% (HPCE)
fluorescence   λex 688 nm; λem 715 nm±5 nm in 0.1 M phosphate pH 7.0
suitability   suitable for fluorescence
storage temp.   2-8°C

Description

Application

for the fluorescence detection of HIS-tagged proteins

Price and Availability

Safety & Documentation

Safety Information

Documents

Certificate of Analysis

Protocols & Articles

Articles

New Products for Detection Applications

Isoelectric focusing (IEF) is a powerful analytical tool for the separation of proteins. In order to ensure the high performance of analysis, isoelectric point (pI) standards are needed. In addition ...
Keywords: Absorption, Applications, Bacterial conjugations, Capillary electrophoresis, Catalysis, Electrophoresis, Gel electrophoresis, Immobilization, Isoelectric focusing, Purification, Separation, Support

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?