05022801 Sigma

Sf21

DOWNLOAD MSDS (PDF)

Properties

Related Categories Cell Culture, ECACC Cell Lines, S-Z More...
biological source   Ovary from insect
growth mode   Adherent
morphology   Spherical
shipped in   dry ice
storage temp.   −196°C

Description

Application

Protein expression using the Baculovirus vector system

General description

For complete product information, please see Sf21

Culture Medium

SF90011 + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 i.e. seeding at 5x10,000 cells/cm2; 28 C. Gently resuspend cells in old culture medium by dislodging the cells by pipetting, or gentle agitation, centrifuge suspension and resuspend cell pellet in fresh medium.

Cell Line Description

Derived from pupal ovarian tissue of spodoptera frugiperda. The cells are highly susceptible to Baculovirus infection and are used in the production of protein products genetically manipulated into Baculovirus vector systems. They were derived at the USDA Insect Pathology Laboratory from a cell line originating from pupal ovarian tissue of the fall army worm, Spodoptera frugiperda. Sf21 cells have greater range in size and have a doubling time of 24 hr compared to 48-72 hr for Sf9 cells.

Price and Availability

NEW! Sigma® Cultureware


Cell Culture SuppliesIntroducing Sigma’s new competitively
priced line of cultureware:

  • . Multiwell Plates
  • . Culture Flasks
  • . Cryogenic Vials & More



Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?