• USA Home
  • 05497 - Albumin blue 580 potassium salt solution

EMAIL THIS PAGE TO A FRIEND
05497 Sigma

Albumin blue 580 potassium salt solution

for fluorescence, in isopropanol

Purchase

Properties

Related Categories Biochemicals and Reagents, Fluorescent Labels, Fluorescent Probes, Labels, Particles and Stains, Fluorescent Stains, Other Fluorescent Labels,
grade   for fluorescence
description   Buffer for the determination of albumin (see Cat. No. 79438)
InChI Key   RDMQASZXPHAORJ-AQCBZIOHSA-N
concentration   in isopropanol
density   0.786 g/mL at 20 °C

Description

Analysis Note

λmax. ∼582 nm, A 1.00 +/-0.05 (0.1 ml/ml 2-propanol)

Application

for the micro-determination of albumin in urine

Features and Benefits

Albumin blue is a stain for the specific and sensitive spectrofluorometric determination of albumin (1-200mg/l) in natural matrices.

Other Notes

Fluorescence assay for albumin

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1219 3 / PGII
WGK Germany 
1
Flash Point(F) 
53.6 °F
Flash Point(C) 
12 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?