• USA Home
  • 06776 - Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) bis(perchlorate) complex

EMAIL THIS PAGE TO A FRIEND
06776 Sigma

Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) bis(perchlorate) complex

BioReagent, suitable for fluorescence, ≥90% (HPCE)

Synonym: Ru(dpp)3-PC

  • CAS Number 75213-31-9

  • Empirical Formula (Hill Notation) C72H48Cl2N6O8Ru

  • Molecular Weight 1297.16

  •  MDL number MFCD03093709

  •  PubChem Substance ID 24845970

Purchase

Properties

Related Categories Biochemicals and Reagents, Luminescent Compounds/Detection, Stains and Dyes More...
InChI Key   MOBZIKDAUJCZDU-UHFFFAOYSA-L
product line   BioReagent
assay   ≥90% (HPCE)
fluorescence   λex 526 nm; λem 631 nm in 0.1 M phosphate pH 7.0
suitability   suitable for fluorescence

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?