• USA Home
  • 09578 - L-(+)-Lactic acid solution

EMAIL THIS PAGE TO A FRIEND
09578 Sigma

L-(+)-Lactic acid solution

purum p.a., 79-81% (L-lactic acid in water, T)

Synonym: (S)-2-Hydroxypropionic acid, Sarcolactic acid

  • CAS Number 79-33-4

  • Empirical Formula (Hill Notation) C3H6O3

  • Molecular Weight 90.08

  •  Beilstein Registry Number 1720251

  •  MDL number MFCD00064266

  •  PubChem Substance ID 329748775

Purchase

Properties

Related Categories Asymmetric Synthesis, Carbohydrates, Carboxylic Acids, Chemical Synthesis, Chiral Building Blocks,
InChI Key   JVTAAEKCZFNVCJ-REOHCLBHSA-N
grade   purum p.a.
assay   79-81% (L-lactic acid in water, T)
impurities   ≤0.001% heavy metals (as Pb)
  ≤1.5% D-lactic acid
ign. residue   ≤0.1% (as SO4)
anion traces   chloride (Cl-): ≤50 mg/kg
  sulfate (SO42-): ≤100 mg/kg
cation traces   Ca: ≤100 mg/kg
  Fe: ≤10 mg/kg

Description

Other Notes

Aqueous solutions of l(+)-Lactate.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
1
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?