EMAIL THIS PAGE TO A FRIEND
10794 Sigma

Antifoam A

silicone polymer based antifoam, emulsion

Purchase

Properties

form   emulsion
concentration   1-50 mg/kg (suppress foaming)
impurities   non-ionic emulsifiers

Description

Physical form

30% aqueous emulsion of silicon polymer

Application

Antifoam A has been used in a study to determine ultra trace arsenic species in water samples by hydride generation atomic absorption spectrometry after cloud point extraction. Antifoam A has also been used in a study to demonstrate a simple method to increase the amount of protein in a culture and to provide insight into how antifoams interact with microbial cell factories.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>213.8 °F
Flash Point(C) 
>101 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?