• USA Home
  • 11337 - Sample buffer according to Lämmli

EMAIL THIS PAGE TO A FRIEND
11337 Sigma

Sample buffer according to Lämmli

BioChemika, for electrophoresis, 2× concentrate

Synonym: Lämmli sample buffer

Purchase

Properties

Related Categories Buffers, Molecular Biology, Protein Electrophoresis, Proteomics, SDS-PAGE More...
grade   for electrophoresis
product line   BioChemika
quality   2× concentrate
composition   bromophenol blue in 0.125 M TRIS HCl, 0.004%
  glycerol, 20%
  2-mercaptoethanol, 10%
  SDS, 4%
pH   ~6.5
suitability   in accordance for electrophoresis test
storage temp.   −20°C

Price and Availability

Safety & Documentation

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?