• USA Home
  • 12826 - 3-(2-Benzoxazolyl)-4-cyanoumbelliferone

EMAIL THIS PAGE TO A FRIEND
12826 Sigma

3-(2-Benzoxazolyl)-4-cyanoumbelliferone

BioReagent, suitable for fluorescence

Synonym: 3-(2-Benzoxazolyl)-4-cyano-7-hydroxycoumarin

  • CAS Number 90146-01-3

  • Empirical Formula (Hill Notation) C17H8N2O4

  • Molecular Weight 304.26

  •  Beilstein Registry Number 6431927

  •  MDL number MFCD01862928

  •  PubChem Substance ID 57647030

Purchase

Properties

Related Categories Biochemicals and Reagents, Detection, Fluorescent Probes, Labels, Particles and Stains, Luminescent Compounds/Detection, Wavelength Index More...
InChI Key   FJBJRHZSCDFLBF-UHFFFAOYSA-N
product line   BioReagent
mp   282-285 °C(lit.)
solubility   DMF: soluble
  DMSO: soluble
  methanol: soluble
fluorescence   λex 505 nm; λem 577 nm in 0.1 M Tris pH 9.0
suitability   suitable for fluorescence

Description

Other Notes

Long wave absorbing and fluorescing pH indicator for the weakly acidic pH range, and enzyme substrate ref. standard

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?