• USA Home
  • 14825 - Bis(4-fluoro-3-nitrophenyl) sulfone

EMAIL THIS PAGE TO A FRIEND
14825 Sigma

Bis(4-fluoro-3-nitrophenyl) sulfone

≥97.0% (HPLC)

Synonym: 4,4′-Difluoro-3,3′-dinitrodiphenyl sulfone, 4-Fluoro-3-nitrophenyl sulfone

  • CAS Number 312-30-1

  • Linear Formula [FC6H3(NO2)]2SO2

  • Molecular Weight 344.25

  •  Beilstein Registry Number 2486439

  •  EC Number 206-224-7

  •  MDL number MFCD00007057

  •  PubChem Substance ID 57647215

Purchase

Properties

InChI Key   KHAWDEWNXJIVCJ-UHFFFAOYSA-N
assay   ≥97.0% (HPLC)
mp   191-194 °C(lit.)
  191-194 °C

Description

Other Notes

Bifunctional reagent for investigating the folding of polypeptide chains; Crosslinking reagent via amino and phenolic hydroxy groups

Application

Bis(4-fluoro-3-nitrophenyl) sulfone has been used to study the high-affinity ATP-binding site on 30S dynein, as well as the staphylococcal protein A in immunoferritin techniques. Bis(4-fluoro-3-nitrophenyl) sulfone has also been used to study ATPase activity and pellet height response of tetrahymena cilia.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
WR4300000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?