• USA Home
  • 15400 - 4-(Boc-aminomethyl)phenyl isothiocyanate

EMAIL THIS PAGE TO A FRIEND
15400 Sigma

4-(Boc-aminomethyl)phenyl isothiocyanate

≥98.0% (HPLC)

Synonym: 4-(N-tert-Butoxycarbonylaminomethyl)phenyl isothiocyanate, tert-Butyl N-(4-isothiocyanatophenylmethyl)carbamate, BAMPITC, N-Boc-4-isothiocyanatobenzylamine

  • CAS Number 89631-74-3

  • Empirical Formula (Hill Notation) C13H16N2O2S

  • Molecular Weight 264.34

  •  Beilstein Registry Number 9557243

  •  MDL number MFCD00077918

  •  PubChem Substance ID 57647410

Purchase

Properties

Related Categories Molecular Biology, Protein Sequencing, Protein Structural Analysis, Proteomics, Reagents for Protein Sequencing More...
InChI Key   FZQIQTXXAATZOS-UHFFFAOYSA-N
assay   ≥98.0% (HPLC)
mp   111-113 °C

Description

Other Notes

Novel protein micro sequencing reagent with a masked amino group which renders the thiohydantoin upon deblocking susceptible to fluorogenic detection.; A 2.5- to 10-fold increase in sensitivity over the most commonly used detection methods was observed (UV-detection at 254 nm)

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




Amplified Detection
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?