EMAIL THIS PAGE TO A FRIEND
15665 Sigma

1,6-Hexanediol

≥99% C6-Dioles basis (GC)

Synonym: Hexamethylene glycol

  • CAS Number 629-11-8

  • Linear Formula HO(CH2)6OH

  • Molecular Weight 118.17

  •  Beilstein Registry Number 1633461

  •  EC Number 211-074-0

  •  MDL number MFCD00002985

Purchase

Properties

vapor pressure   0.53 mmHg ( 20 °C)
InChI Key   XXMIOPMDWAUFGU-UHFFFAOYSA-N
assay   ≥99% C6-Dioles basis (GC)
autoignition temp.   608 °F
expl. lim.   16 %
bp   250 °C(lit.)
mp   38-42 °C(lit.)
  38-42 °C

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
MO2100000
Flash Point(F) 
215.6 °F
Flash Point(C) 
102 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?