• USA Home
  • 16569 - 4-Bromo-calcium Ionophore A23187

EMAIL THIS PAGE TO A FRIEND
16569 Sigma

4-Bromo-calcium Ionophore A23187

≥95.0% (N)

Synonym: 4-Bromo-A23187, 4-Bromo-calcimycin

  • CAS Number 76455-48-6

  • Empirical Formula (Hill Notation) C29H36BrN3O6

  • Molecular Weight 602.52

  •  MDL number MFCD00077667

  •  PubChem Substance ID 57647578

Purchase

Properties

Related Categories Antibiotics, Antibiotics A to Z, Antibiotics A-F, Biochemicals and Reagents, Interferes with Cell Membrane Permeability (Ionophores),
material   faint yellow to yellow
InChI Key   LDXQFZZGVTWFCF-FHYGWRBKSA-N
assay   ≥95.0% (N)
form   powder or crystals
solubility   ethanol: soluble20 mg/mL, clear, yellow
Mode of action   cell membrane | interferes
storage temp.   2-8°C

Description

Application

Nonfluorescent Ca2+ ionophore that can be used to calibrate the quantitation of cytoplasmic free Ca2+ with fluorescent probes.

Bromo-A23187, a nonfluorescent ionophore, has been used in photolysis of a caged-Ca2+ compound and in [Ca2+]i elevation protocols. It can be used to calibrate the quantitation of cytoplasmic free Ca2+ with fluorescent probes.

Other Notes

In biological environment, bromo-A23187 has a higher selectivity for Ca2+ over Mg2+ than A23187

Biochem/physiol Actions

Ionophores disrupt transmembrane ion concentration gradients which are required for proper functioning and survival of microorganisms. An ionophore, such as Bromo-A23187, increases the permeability of biological membranes to calcium.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?