Synonym: Lipase, Candida antarctica, cross-linked enzyme aggregate
| Related Categories | 3.1.x.x Acting on esters, 3.x.x.x Hydrolases, Biochemicals and Reagents, Chemical Synthesis, Enzyme Class Index, |
| storage temp. | 2-8°C |
CLEA (Cross-Linked Enzyme Aggregate) technology1
1 U corresponds to the amount of enzyme which hydrolyzes 1 μmol acetic acid per minute at pH 7.4 and 40°C (triacetin, Fluka No. 90240 as substrate)
Customers Also Viewed
≥1.5 U/mg
≥5,000 U/g, recombinant, expressed in Aspergillus niger
≥1800 U/g
powder, beige, ~9 U/mg
≥1.0 U/mg
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?