• USA Home
  • 17082 - Fura PE 3 hexapotassium salt

EMAIL THIS PAGE TO A FRIEND
17082 Sigma

Fura PE 3 hexapotassium salt

for fluorescence, ≥90% (TLC)

  • Empirical Formula (Hill Notation) C37H33K6N5O17

  • Molecular Weight 1054.27

  •  MDL number MFCD01866713

  •  PubChem Substance ID 24850325

Purchase

Properties

Related Categories Biochemicals and Reagents, Calcium Indicators, Chelators and Ion Probes, Fluorescent Indicators and Probes, Fluorescent Probes, Labels, Particles and Stains,
grade   for fluorescence
InChI Key   KSBGDEQSGMNOBS-UHFFFAOYSA-H
assay   ≥90% (TLC)
solubility   H2O: soluble
fluorescence   λex 335 nm; λem 495 nm in 0.1 M Tris pH 8.0
storage temp.   −20°C

Description

Features and Benefits

Off white solid. Fluorescent calcium probe with similar applications and fluorescent properties as Fura-2. Fura-PE3 is retained intracellularly longer than Fura-2.

Other Notes

Fluorescent calcium probe; abs.: λmax 364 nm; em.: λmax 502 nm

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?