• USA Home
  • 17283 - 2′,7′-Bis(3-carboxypropyl)-5(6)-carboxyfluorescein

EMAIL THIS PAGE TO A FRIEND
17283 Sigma

2′,7′-Bis(3-carboxypropyl)-5(6)-carboxyfluorescein

BioReagent, suitable for fluorescence, ≥95.0% (HPCE)

Synonym: BCPCF

  • CAS Number 202872-98-8

  • Empirical Formula (Hill Notation) C29H24O11

  • Molecular Weight 548.49

  •  PubChem Substance ID 329751300

Purchase

Properties

Related Categories Alphabetical, Biochemicals and Reagents, Fluorescent Indicators and Probes, Fluorescent Probes, Labels, Particles and Stains, pH Sensitive Probes and Indicators More...
InChI Key   YKFRFHRHRHPYNI-UHFFFAOYSA-N
product line   BioReagent
assay   ≥95.0% (HPCE)
solubility   H2O: soluble (pH ≥ 6)
fluorescence   λex 480 nm; λem 527 nm in 0.1 M Tris pH 9.0
suitability   suitable for fluorescence
storage temp.   2-8°C

Description

Other Notes

Improved analogue of the fluoresent indicator for cytoplamsic pH BCECF

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Sensor Dyes

In 1998 R.F. Furchgott, L.J. Ignarro, and F. Murad received the Nobel Prize in Physiology and Medicine for their discoveries concerning nitric oxide as a signaling molecule in the cardiovascular syst...
Monika Baeumle
BioFiles 2011, 6.3, 23.
Keywords: Applications, Biochemistry, Biological processes, Cancer, Cardiovascular, Central Nervous System, Diseases, Extinction coefficient, Indicators, Inflammation, Methods, Microscopy, Neurotransmission, Sol-gel, Type

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?