• USA Home
  • 17861 - Biotin-CryptoFluor Crimson
EMAIL THIS PAGE TO A FRIEND
17861 Sigma
Biotin-CryptoFluor Crimson
BioReagent, suitable for fluorescence
Purchase
Description
General description
The CryptoFluor dyes are phycobiliproteins isolated from cryptomonad algae that provide additional spectral characteristics to complement the more common phycobiliproteins (e.g., R-PE and APC) as well as providing much smaller molecular weights (around 40,000 Daltons). These characteristics are meant to provide additional functionality to the phycobiliprotein pigments when it comes to multiplexing assays or to getting dyes into the cell.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
One vial contents 0.5 mg of conjugate and ~11 mg additives (see reconstitution)
Reconstitution
Reconstitute to 250 μl with distilled or deionized water (the solution will consist of 100 mM sodium phosphate, pH 7.2, 50 mM sucrose, 0.05% sodium azide, 150 mM sodium chloride, 0.1% bovine serum albumin and 2 mg/ml pigment. Do not freeze, store at 4 °C in the dark.
Legal Information
CryptoFluor is a trademark of Martek Biosciences Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?