• USA Home
  • 1802 - Naphthol AS-BI Phosphoric acid solution

EMAIL THIS PAGE TO A FRIEND
1802 Sigma

Naphthol AS-BI Phosphoric acid solution

Purchase

Properties

Related Categories Cell Biology, Cytochemistry Products, Hematology and Histology More...
InChI Key   HUXIAXQSTATULQ-UHFFFAOYSA-N
Quality Level   GMP-IVD
shelf life   Expiry date on the label.
IVD   for in vitro diagnostic use
shipped in   dry ice
storage temp.   −20°C

Description

Components

Napthol AS-BI phosphoric Acid, 4 g/L, in methanol solution with stabilizers.

Application

For use in the Sigma Lymphocyte Enzyme (Acid Phosphatase) Procedure, No. 181A

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

Signal word 
Danger
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
2
Flash Point(F) 
96.8 °F
Flash Point(C) 
36 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?