• USA Home
  • 18115 - Malic Dehydrogenase, E. coli, recombinant from E. coli

EMAIL THIS PAGE TO A FRIEND
18115 Sigma

Malic Dehydrogenase, E. coli, recombinant from E. coli

≥300 units/mL

Synonym: MDH

Purchase

Properties

storage temp.   −20°C

Description

Unit Definition

1 U corresponds to the amount of enzyme which converts 1 μmol L-malate to pyruvate and CO2 per minute at pH 7.5 and 30°C (NADP as cofactor)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?