• USA Home
  • 181A - Acid Phosphatase, Lymphocyte

EMAIL THIS PAGE TO A FRIEND
181A Sigma

Acid Phosphatase, Lymphocyte

Purchase

Properties

Related Categories Cell Biology, Cytochemistry Products, Hematology and Histology More...
Quality Level   GMP-IVD
shelf life   Expiry date on the label.
IVD   for in vitro diagnostic use
shipped in   wet ice
storage temp.   2-8°C

Description

Diagnostic Implications

Demonstration of lymphocyte acid phosphatase (focal staining pattern) in cytocentrifuge or buffy coat preparations from blood, bone marrow film and tissue touch preparations typifies T-lymphocytes.

Price and Availability

Kit component also available separately

Description

Product #

Add to Cart

Acetate Solution (386-3) 50 mL SDS    
Citrate Solution (91-5) 50 mL SDS    
Fast Garnet GBC Base Solution (387-2) 10 mL SDS    
Methylene Blue Solution (180-8) 50 mL SDS    
Naphthol AS-BI Phosphoric Acid Solution (180-2) 50 mL SDS    
Sodium Nitrite Solution (91-4) 10 mL SDS    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3316 9
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

915 Citrate Solution, pH 3.6±0.1 (25 °C), 27 mM
3863 Acetate solution, 2.5 M, pH 5.2
3872 Fast Garnet GBC Base solution
1802 Naphthol AS-BI Phosphoric acid solution
1808 Methylene Blue solution, 1.4 % (w/v) in 95% ethanol
R8382 Reagent Alcohol, used for histology tissue preparation
914 Sodium nitrite solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?