• USA Home
  • 20777 - Butyrylcholinesterase from equine serum

EMAIL THIS PAGE TO A FRIEND
20777 Sigma

Butyrylcholinesterase from equine serum

lyophilized, powder, light brown, 7-16 U/mg

Synonym: Acylcholine acyl-hydrolase, Choline esterase, butyryl, Pseudocholinesterase

Purchase

Properties

Related Categories 3.1.x.x Acting on esters, 3.x.x.x Hydrolases, Biochemicals and Reagents, Cell Biology, Enzyme Class Index,
form   powder
quality   lyophilized
mol wt   Mr ~440000
color   light brown
storage temp.   −20°C

Description

Other Notes

Comparison of butyrylcholinesterase and acetylcholinesterase, review; Applications in (selective) hydrolysis/condensation of carboxylic ester bonds.

Sales restrictions may apply

Unit Definition

1 U corresponds to the amount of enzyme which hydrolyzes 1 μmol butyrylcholine per minute at pH 8.0 and 37 °C.

Price and Availability


Available in ELITE Grade
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?