• USA Home
  • 21807 - Carbonic Anhydrase Isozyme II from bovine erythrocytes

EMAIL THIS PAGE TO A FRIEND
21807 Sigma

Carbonic Anhydrase Isozyme II from bovine erythrocytes

powder, white, ~1800 U/mg

Synonym: Carbonate Dehydratase, Carbonate Hydrolyase, Carbonic Anhydrase II

Purchase

Description

Unit Definition

1 U corresponds to the amount of enzyme which causes the pH of a 20 mM TRIS Buffer to drop from 8.3 to 6.3 per minute at 0°C (Wilbur-Andersen U)

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C2522 Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥3,000 W-A units/mg protein

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?