• USA Home
  • 21851 - Carbonate/Bicarbonate buffer substance

EMAIL THIS PAGE TO A FRIEND
21851 Sigma

Carbonate/Bicarbonate buffer substance

for enzyme immunoassay, capsule

Synonym: Buffer substance from carbonate and bicarbonate

Purchase

Properties

Related Categories Biochemicals and Reagents, Biological Buffers, Buffer Capsules, Buffer Convenience Packaging, Buffers A to Z More...
grade   for enzyme immunoassay
form   capsule

Description

General description

The capsules are to be opened first, then the contents should be droppped into the deionized water. The capsules themselves are not to be placed in the water.

Packaging

Package of 50 capsules

Reconstitution

1 capsule dissolved in 100 mL water gives a 0.05 M solution pH 9.6 (25°C)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?