• USA Home
  • 25116 - (Chloromethyl)dimethylphenylsilane

EMAIL THIS PAGE TO A FRIEND
25116 Sigma

(Chloromethyl)dimethylphenylsilane

purum, ≥98.0% (GC)

  • CAS Number 1833-51-8

  • Linear Formula C6H5Si(CH3)2CH2Cl

  • Molecular Weight 184.74

  •  Beilstein Registry Number 2935991

  •  EC Number 217-392-6

  •  MDL number MFCD00040858

  •  PubChem Substance ID 329752777

Purchase

Properties

InChI Key   RJCTVFQQNCNBHG-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.520(lit.)
  n20/D 1.520
bp   106-107 °C/15 mmHg(lit.)
density   1.024 g/mL at 25 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
195.8 °F
Flash Point(C) 
91 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

347094 (Chloromethyl)dimethylphenylsilane, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?