• USA Home
  • 25925 - 1-(2-Chlorophenyl)-1-(4-chlorophenyl)-2,2-dichloroethane

EMAIL THIS PAGE TO A FRIEND
25925 Sigma

1-(2-Chlorophenyl)-1-(4-chlorophenyl)-2,2-dichloroethane

for diagnostic uses (cancer investigation), ≥98% (HPLC)

Synonym: (2,4′-Dichlorodiphenyl)dichloroethane, o,p′-DDD, Mitotane

  • CAS Number 53-19-0

  • Linear Formula (ClC6H4)2CHCHCl2

  • Molecular Weight 320.04

  •  Beilstein Registry Number 2056007

  •  EC Number 200-166-6

  •  MDL number MFCD00000850

  •  PubChem Substance ID 24855696

Purchase

Properties

grade   for diagnostic uses (cancer investigation)
InChI Key   JWBOIMRXGHLCPP-UHFFFAOYSA-N
assay   ≥98% (HPLC)
mp   76-78 °C
Gene Information   rat ... Ar(24208)

Description

Packaging

1 g in glass bottle

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
KH7880000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?