• USA Home
  • 29467 - Cyclohexylmethyl β-D-maltoside

EMAIL THIS PAGE TO A FRIEND
29467 Sigma

Cyclohexylmethyl β-D-maltoside

≥99.0% (TLC)

Synonym: CYMAL®-1

Purchase

Properties

Related Categories Biochemicals and Reagents, Detergents, Detergents A to M, Detergents by Applications, Electrophoresis Chromatography,
description   non-ionic
InChI Key   BBUKPWNPDDHXDR-CRINDFDSSA-N
assay   ≥99.0% (TLC)
CMC   340

Description

Application

Detergent for the purification and crystallization of membrane-bound proteins in native structure. It has a higher CMC (critical micelle concentration) and is more hydrophobe than the corresponding linear chain analogue (C19): M. Ju, Z.E. Rassi, J. Liqu. Chromatogr. Relat. Technol., 23, 35 (2000)

Legal Information

No license is conveyed with the purchase of this product.

CYMAL is a registered trademark of Affymetrix, Inc.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?