• USA Home
  • 304557 - Anthranilic acid-ring-UL-14C

EMAIL THIS PAGE TO A FRIEND
304557 Sigma

Anthranilic acid-ring-UL-14C

solid

Synonym: 2-Aminobenzoic-ring-ul-14C acid

Purchase

Properties

InChI Key   RWZYAGGXGHYGMB-UHFFFAOYSA-N
form   solid
extent of labeling   1-25 mCi per mmol
storage temp.   2-8°C

Description

Packaging

screw cap bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?