• USA Home
  • 304654 - 1,4-Dichlorobenzene-UL-14C

EMAIL THIS PAGE TO A FRIEND
304654 Sigma

1,4-Dichlorobenzene-UL-14C

neat

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Labeled Pesticide Standards, Pesticides & Metabolites Standards
InChI Key   OCJBOOLMMGQPQU-UHFFFAOYSA-N
form   neat
extent of labeling   5-15 mCi per mmol
storage temp.   2-8°C
Gene Information   human ... CYP2A6(1548)
mouse ... Cyp2a5(13087)

Description

Packaging

Sealed ampule

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
3
Flash Point(F) 
151 °F
Flash Point(C) 
66 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?