• USA Home
  • 311588 - Sodium acetate-1-14C

EMAIL THIS PAGE TO A FRIEND
311588 Sigma

Sodium acetate-1-14C

≥98% radiochemical purity basis (HPLC), aqueous solution

Synonym: Acetic acid-1-14C sodium salt

Purchase

Properties

InChI Key   VMHLLURERBWHNL-UOEUYCPOSA-M
assay   ≥98% radiochemical purity basis (HPLC)
form   aqueous solution
extent of labeling   40-60 mCi per mmol
mass shift   M+2
storage temp.   2-8°C

Description

Packaging

Combi-vial

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?