EMAIL THIS PAGE TO A FRIEND
314722 Sigma

Benzil-ring-UL-14C

Purchase

Properties

storage temp.   2-8°C

Description

Other Notes

deleted item

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?