• USA Home
  • 322032 - Bromoacetic acid-carboxy-14C

EMAIL THIS PAGE TO A FRIEND
322032 Sigma

Bromoacetic acid-carboxy-14C

crystalline

Purchase

Properties

InChI Key   KDPAWGWELVVRCH-HQMMCQRPSA-N
form   crystalline
extent of labeling   10-30 mCi per mmol
storage temp.   2-8°C

Description

Packaging

screw cap bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?