• USA Home
  • 33170 - 1,8-Diaminonaphthalene

EMAIL THIS PAGE TO A FRIEND
33170 Sigma

1,8-Diaminonaphthalene

for fluorescence, ≥98.0% (HPLC/NT)

Synonym: 1,8-Naphthalenediamine

  • CAS Number 479-27-6

  • Linear Formula C10H6(NH2)2

  • Molecular Weight 158.20

  •  Beilstein Registry Number 742841

  •  EC Number 207-529-8

  •  MDL number MFCD00004033

  •  PubChem Substance ID 57648771

Purchase

Properties

Related Categories Biochemicals and Reagents, Detection, Fluorescent Probes, Labels, Particles and Stains, Luminescent Compounds/Detection, Wavelength Index More...
grade   for fluorescence
InChI Key   YFOOEYJGMMJJLS-UHFFFAOYSA-N
assay   ≥98.0% (HPLC/NT)
bp   205 °C/12 mmHg(lit.)
mp   60-65 °C(lit.)
  63-67 °C
solubility   methanol: soluble
fluorescence   λex 356 nm; λem 392 nm in cyclohexane

Description

Caution

may discolor to brown on storage

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?