• USA Home
  • 33790 - 1,3-Diphenyl-2-propanone

EMAIL THIS PAGE TO A FRIEND
33790 Sigma

1,3-Diphenyl-2-propanone

purum, ≥98.0% (GC)

Synonym: 1,3-Diphenylacetone, Dibenzyl ketone

  • CAS Number 102-04-5

  • Linear Formula (C6H5CH2)2CO

  • Molecular Weight 210.27

  •  Beilstein Registry Number 974767

  •  EC Number 203-000-0

  •  MDL number MFCD00004795

  •  PubChem Substance ID 24860503

Purchase

Properties

Related Categories Building Blocks, C15 to C38, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   YFKBXYGUSOXJGS-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
bp   330 °C(lit.)
mp   32-34 °C(lit.)
  32-34 °C
Gene Information   human ... EPHX2(2053)
mouse ... Ephx2(13850)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

1,3-Diphenyl-2-propanone was used in the preparation of isomeric pyranones via condensation with propionic acid.

General description

1,3-Diphenyl-2-propanone is metabolite of α-benzyl-N-methylphenethylamine (BNMPA), an impurity of illicit methamphetamine synthesis. 1,3-Diphenyl-2-propanone undergoes α-ethylation with ethyl iodide and sodium ethoxide to yield 1,3-dipenyl-propene. 1,3-Diphenyl-2-propanone undergoes condensation with acetic acid to yield 2,6-dimethyl-3,5-diphenylpyran-4-one.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
EC2030000
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?