• USA Home
  • 3392B - BIOEAZE® Ethyl Vinyl Acetate (EVA) Bags

EMAIL THIS PAGE TO A FRIEND
3392B Sigma

BIOEAZE® Ethyl Vinyl Acetate (EVA) Bags

volume 1 L

Synonym: 0.5-1 L Ethyl Vinyl Acetate (EVA) sterile bag with BioQuate Disposable Aseptic Connector (DAC)

Purchase

Properties

sterility   sterile; γ-irradiated
packaging   pkg of 1
volume   1 L

Description

Features and Benefits

BIOEAZE bioprocess bags are specifically designed to provide strong seals, extraordinary robustness, superior flex, crack and pin-hole resistance, plus desirable gas and moisture barrier performance.

BIOEAZE Ethyl Vinyl Acetate (EVA) bags are constructed from a single-web, medical grade laminate of four different films, with a fluid contact surface of EVA, with two layers of low-density polyethylene (LDPE) enveloping a layer of ethyl vinyl alcohol (EVOH).

General description

Designed for use with the ReadyMate Disposable Aseptic Connector (DAC) or Terumo Aseptic Connecting Device (ACD)

Configured with:
Port P1: 1 x 6" length of 1/4" x 7/16" Clear 082 Clear C-Flex® Tubing with Pinch Clamp and Disposable Aseptic Connector (DAC)
Port P2: 1 x 1 ft. length of Opaque 001 Size 16 C-Flex® Tubing with Pinch Clamp and Plug
Port P3: 1 Spike Port

Packaging

Double Polybagged

Legal Information

BIOEAZE is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?