EMAIL THIS PAGE TO A FRIEND
34431 Sigma

CBinD L

suspension in PBS

Synonym: Protein L, immobilized on cellulose

Purchase

Properties

Related Categories Affinity Chromatography, Antibodies, Antibody Purification and Characterization, Molecular Biology, Protein A, G and L Resins,
quality   suspension in PBS
contains   ~20% ethanol as stabilizer
capacity   ≥5 mg/mL binding capacity (human IgG)
storage temp.   2-8°C

Description

General description

Immobilized rCBD-Protein L on cellulose beads (covalently cross-linked). Designed for quick and efficient purification of immunoglobulins (Ig) and Ig fragments (Fab and scFr). ~2.5 mg rCBD-Protein L per mL cellulose suspension.

Other Notes

An immobilized rCBD-Protein L matrix (CBD: Cellulose Binding Proteins) immobilized on cellulose beads. Direct purification of monoclonal antibodies from media supplemented with FCS

Packaging

Pack sizes comply with the amount of suspension

Legal Information

CBinD is a trademark of CBD Technologies, Inc.

Price and Availability


Antibody Explorer

Explore with Confidence

Amplified Detection
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?