• USA Home
  • 34714 - AQUANAL RH mixed indicator

EMAIL THIS PAGE TO A FRIEND
34714 Sigma

AQUANAL RH mixed indicator

for the determination of residual water hardness

Synonym: Mixed Indicator RH

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Kits/Cases, Potable- & Waste-Water Testing
quality   for the determination of residual water hardness
impurities   sodium carbonate

Description

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?