• USA Home
  • 36190 - 2,6-Dichlorophenolindophenyl acetate

EMAIL THIS PAGE TO A FRIEND
36190 Sigma

2,6-Dichlorophenolindophenyl acetate

≥97.0% (CHN)

Synonym: DIPA

Purchase

Description

Other Notes

Substrate employed in the spectrophotometric enzymatic determination of organophosphorus compounds

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?