EMAIL THIS PAGE TO A FRIEND
36660 Sigma

Dieldrin solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

  • CAS Number 60-57-1

  • Empirical Formula (Hill Notation) C12H8Cl6O

  • Molecular Weight 380.91

  •  Beilstein Registry Number 91396

  •  MDL number MFCD00135591

  •  PubChem Substance ID 329755435

Purchase

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1648 3 / PGII
WGK Germany 
2
Flash Point(F) 
35.6 °F
Flash Point(C) 
2 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of Agriculture Pesticides in Orange (Spiked) on SLB®-5ms after QuEChERS Cleanup using Primary-Secondary Amine (PSA)

From our library of Articles, Sigma-Aldrich presents GC Analysis of Agriculture Pesticides in Orange (Spiked) on SLB®-5ms after QuEChERS Cleanup using Primary-Secondary Amine (PSA)
Keywords: Chromatography, Gas chromatography, Pesticides, Purification, Sample preparations, Solid phase extractions

GC Analysis of Multiple Pesticides on SLB®-5ms after SPME using 60 μm Carbowax® Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Multiple Pesticides on SLB®-5ms after SPME using 60 μm Carbowax® Fiber
Keywords: Agriculture, Chromatography, Gas chromatography, Metabolites, Pesticides, Purification, Solid phase microextractions

GC Analysis of Organochlorine Pesticides in Water on SPB®-5 after SPME using 100 μm PDMS Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Organochlorine Pesticides in Water on SPB®-5 after SPME using 100 μm PDMS Fiber
Keywords: Chromatography, Electron capture detector, Environmental, Gas chromatography, Metabolites, Pesticides, Purification, Size-exclusion chromatography, Solid phase microextractions

GC Analysis of a 20-Component Organochlorine Pesticide Mix in Water on a 5% Phenyl Column after SPE using Supelclean™ ENVI™-8

From our library of Articles, Sigma-Aldrich presents GC Analysis of a 20-Component Organochlorine Pesticide Mix in Water on a 5% Phenyl Column after SPE using Supelclean™ ENVI™-8
Keywords: Chromatography, Electron capture detector, Gas chromatography, Purification, Sample preparations, Size-exclusion chromatography, Solid phase extractions

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on Equity®-1701 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on Equity®-1701 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Keywords: Chromatography, Electron capture detector, Gas chromatography, Pesticides, Purification, Size-exclusion chromatography

US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SLB®-5ms (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SLB®-5ms (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Keywords: Chromatography, Electron capture detector, Gas chromatography, Pesticides, Purification, Size-exclusion chromatography

US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SLB®-5ms
Keywords: Chromatography, Electron capture detector, Gas chromatography, Pesticides, Purification

US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SPB®-608

From our library of Articles, Sigma-Aldrich presents US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SPB®-608
Keywords: Chromatography, Electron capture detector, Gas chromatography, Pesticides, Purification

US EPA Method 625: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 625: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8081: GC Analysis of Organochlorine Pesticides on Equity®-5

From our library of Articles, Sigma-Aldrich presents US EPA Method 8081: GC Analysis of Organochlorine Pesticides on Equity®-5
Keywords: Chromatography, Electron capture detector, Environmental, Gas chromatography, Pesticides, Purification, Size-exclusion chromatography

US EPA Method TO-10/IP-8 and ASTM® D4861: GC Analysis of Pesticides on SPB®-608 after Collection/Desorption using ORBO™-1000

From our library of Articles, Sigma-Aldrich presents US EPA Method TO-10/IP-8 and ASTM® D4861: GC Analysis of Pesticides on SPB®-608 after Collection/Desorption using ORBO™-1000
Keywords: Air Monitoring, Chromatography, Electron capture detector, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?