EMAIL THIS PAGE TO A FRIEND
36788 Sigma

Dimefuron

PESTANAL®, analytical standard

  • CAS Number 34205-21-5

  • Empirical Formula (Hill Notation) C15H19ClN4O3

  • Molecular Weight 338.79

  •  Beilstein Registry Number 628305

  •  EC Number 251-879-4

  •  MDL number MFCD00144130

  •  PubChem Substance ID 329755510

Purchase

Properties

Related Categories A-D, All Herbicide Standards, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   DHWRNDJOGMTCPB-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
format   neat

Description

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
YS4000000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?